siRNA Details
Gene Symbol: PUM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014878.4 | NP_055693.4 | 13 | 1322 | ||
GenBank mRNA | AF490254.1 | - | 10 | 806 | ||
AK225953.1 | - | 13 | 1287 | |||
AK292169.1 | - | 13 | 1265 | |||
AK315531.1 | - | 13 | 1301 | |||
AL832239.1 | - | 13 | 1288 | |||
AL832245.1 | - | 13 | 1279 | |||
BC016137.2 | - | 12 | 1226 | 3908516 | ||
CR456957.1 | - | 10 | 806 | |||
D13645.1 | - | 12 | 1224 |
