siRNA Details
Gene Symbol: Oxct1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127580.1 | NP_001121052.1 | 16 | 1545 | ||
GenBank mRNA | AY724483.1 | - | 16 | 1499 | ||
BC166478.1 | - | 16 | 1545 | 9102243 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Oxct1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127580.1 | NP_001121052.1 | 16 | 1545 | ||
GenBank mRNA | AY724483.1 | - | 16 | 1499 | ||
BC166478.1 | - | 16 | 1545 | 9102243 |