siRNA Details
Gene Symbol: Harbi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113793.3 | NP_001107265.2 | 3 | 994 | ||
XM_006234587.2 | XP_006234649.1 | 3 | 1641 | |||
XM_006234588.2 | XP_006234650.1 | 3 | 1046 | |||
GenBank mRNA | BC158734.1 | - | 3 | 994 | 9098654 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
