siRNA Details
Gene Symbol: Wdr74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006230999.2 | XP_006231061.1 | 9, 10 | 966 | ||
XM_006231000.2 | XP_006231062.1 | 9, 10 | 834 | |||
GenBank mRNA | BC158834.1 | - | 9, 10 | 928 | 7383724 |

More Information
Other Species Targeted

Gene Ontology Categories:
