siRNA Details
Gene Symbol: Senp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768807.1 | XP_008767029.1 | 7 | 2194 | ||
XM_008768808.1 | XP_008767030.1 | 7 | 2190 | |||
XM_221369.8 | XP_221369.2 | 7 | 2193 | |||
GenBank mRNA | FQ233817.1 | - | 1 | 40 |
Gene Symbol: LOC103689978 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006248475.2 | XP_006248537.1 | 7 | 2193 | ||
XM_008768779.1 | XP_008767001.1 | 7 | 2194 | |||
XM_008768780.1 | XP_008767002.1 | 7 | 2190 |
Gene Symbol: RGD1564247 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749241.3 | XP_003749289.1 | 1 | 2178 |
