siRNA Details
Gene Symbol: Polr3gl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109571.1 | NP_001103041.1 | 6, 7 | 569 | ||
XM_008761331.1 | XP_008759553.1 | 6, 7 | 652 | |||
GenBank mRNA | BC168229.1 | - | 6, 7 | 528 | 7377062 | |
FQ214367.1 | - | 6, 7 | 555 |
