siRNA Details
Gene Symbol: LOC690326 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109578.1 | NP_001103048.1 | 5 | 298 | ||
XM_006237459.1 | XP_006237521.1 | 6 | 472 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
