siRNA Details
Gene Symbol: Cyp3a73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006248850.2 | XP_006248912.1 | 6 | 1645 | ||
XM_008769027.1 | XP_008767249.1 | 6 | 1682 | |||
XM_008769028.1 | XP_008767250.1 | 5 | 1074 | |||
XM_008769029.1 | XP_008767251.1 | 6 | 751 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
