siRNA Details
Gene Symbol: LOC690344 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168661.1 | NP_001162132.1 | 2 | 558 | ||
GenBank mRNA | FM119083.1 | - | 2 | 558 |

More Information
Other Species Targeted

