siRNA Details
Gene Symbol: Lbhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109581.2 | NP_001103051.2 | 4 | 342 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lbhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109581.2 | NP_001103051.2 | 4 | 342 |