siRNA Details
Gene Symbol: Lyrm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126096.1 | NP_001119568.1 | 2, 3 | 211 | ||
GenBank mRNA | BC166578.1 | - | 2, 3 | 211 | 7379886 |

More Information
Other Species Targeted

