siRNA Details
Gene Symbol: Lyrm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126096.1 | NP_001119568.1 | 2 | 135 | ||
GenBank mRNA | BC166578.1 | - | 2 | 135 | 7379886 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
