siRNA Details
Gene Symbol: Bpifa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109588.1 | NP_001103058.1 | 5 | 803 | ||
XM_006235362.2 | XP_006235424.1 | 5 | 821 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
