siRNA Details
Gene Symbol: Taf1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191107.1 | NP_001178036.1 | 4 | 251 | ||
XM_006239995.2 | XP_006240057.1 | 4 | 500 |

More Information
Other Species Targeted

Gene Ontology Categories:
