siRNA Details
Gene Symbol: LOC102548728 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758825.1 | XP_008757047.1 | 3 | 792 | ||
XM_008758827.1 | XP_008757049.1 | 3 | 443 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
