siRNA Details
Gene Symbol: LOC102548728 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758825.1 | XP_008757047.1 | 3 | 796 | ||
XM_008758827.1 | XP_008757049.1 | 3 | 447 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC102548728 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758825.1 | XP_008757047.1 | 3 | 796 | ||
XM_008758827.1 | XP_008757049.1 | 3 | 447 |