siRNA Details
Gene Symbol: Tas1r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271266.1 | NP_001258195.1 | 6 | 2451 | ||
GenBank mRNA | AF127390.1 | - | 6 | 2451 |

More Information
Other Species Targeted

Set Membership: |
![]() |
