siRNA Details
Gene Symbol: LOC102546467 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765531.1 | XP_008763753.1 | 4 | 705 |
Gene Symbol: Oc90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134765.1 | NP_001128237.1 | 4 | 339 | ||
XM_008765519.1 | XP_008763741.1 | 17 | 1856 |
