siRNA Details
Gene Symbol: Fbxo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109606.1 | NP_001103076.1 | 9 | 1022 | ||
XM_006234656.2 | XP_006234718.1 | 9 | 1047 | |||
XM_006234657.2 | XP_006234719.1 | 9 | 1047 | |||
GenBank mRNA | FQ210238.1 | - | 2 | 165 |
