siRNA Details
Gene Symbol: Hint1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109607.1 | NP_001103077.1 | 1, 2 | 171 | ||
GenBank mRNA | BC168732.1 | - | 1, 2 | 184 | 6888300 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
