siRNA Details
Gene Symbol: Vom2r19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099492.1 | NP_001092962.1 | 2 | 242 |
Gene Symbol: LOC100910798 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750426.3 | XP_003750474.2 | 242 |
Gene Symbol: LOC103689981 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758849.1 | XP_008757071.1 | 242 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
