siRNA Details
Gene Symbol: Med12l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169130.1 | NP_001162601.1 | 13 | 2066 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Med12l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169130.1 | NP_001162601.1 | 13 | 2066 |