siRNA Details
Gene Symbol: Vom2r22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099656.1 | NP_001093126.1 | 2528 | |||
XM_003750425.1 | XP_003750473.1 | 6 | 2528 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vom2r22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099656.1 | NP_001093126.1 | 2528 | |||
XM_003750425.1 | XP_003750473.1 | 6 | 2528 |