siRNA Details
Gene Symbol: LOC100911224 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228161.2 | XP_006228223.1 | 4 | 602 |
Gene Symbol: Zik1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228165.2 | XP_006228227.1 | 4 | 729 |
