siRNA Details
Gene Symbol: Polr2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109614.1 | NP_001103084.1 | 3 | 489 | ||
XM_006241009.2 | XP_006241071.1 | 3 | 618 | |||
XM_006241010.1 | XP_006241072.1 | 3 | 509 | |||
XM_006241011.1 | XP_006241073.1 | 3 | 536 | |||
GenBank mRNA | BC158787.1 | - | 3 | 447 | 7304442 | |
FQ219797.1 | - | 3 | 421 | |||
FQ220392.1 | - | 3 | 419 | |||
FQ229401.1 | - | 3 | 536 | |||
FQ230290.1 | - | 3 | 424 | |||
FQ234550.1 | - | 3 | 469 |
