siRNA Details
Gene Symbol: LOC685976 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766820.1 | XP_008765042.1 | 5 | 1745 | ||
XM_008766821.1 | XP_008765043.1 | 5 | 1745 | |||
XM_008766822.1 | XP_008765044.1 | 5 | 641 | |||
XM_008766824.1 | XP_008765046.1 | 5 | 411 |
Gene Symbol: LOC103690960 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758721.1 | XP_008756943.1 | 527 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
