siRNA Details
Gene Symbol: LOC685976 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766820.1 | XP_008765042.1 | 5, 6 | 1794 | ||
XM_008766821.1 | XP_008765043.1 | 5, 6 | 1794 | |||
XM_008766822.1 | XP_008765044.1 | 5, 6 | 690 | |||
XM_008766824.1 | XP_008765046.1 | 5, 6 | 460 |
Gene Symbol: LOC102550773 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006232463.2 | XP_006232525.1 | 119 |
Gene Symbol: LOC103690960 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758721.1 | XP_008756943.1 | 576 |
Gene Symbol: LOC304239 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC166500.1 | - | 292 |
Gene Symbol: LOC680627 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771013.1 | XP_008769235.1 | 5, 6 | 778 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
