siRNA Details
Gene Symbol: Fancg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763655.1 | XP_008761877.1 | 14 | 2416 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fancg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763655.1 | XP_008761877.1 | 14 | 2416 |