siRNA Details
Gene Symbol: Capza1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109625.2 | NP_001103095.2 | 10 | 1107 | ||
GenBank mRNA | BC166464.1 | - | 10 | 822 | 9030793 | |
CB557114.1 | - | 6 | 576 |

More Information
Other Species Targeted

Gene Ontology Categories:
