siRNA Details
Gene Symbol: Lypd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109632.1 | NP_001103102.1 | 5 | 455 | ||
XM_006246402.2 | XP_006246464.1 | 4 | 573 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
