siRNA Details
Gene Symbol: Pglyrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241093.2 | XP_006241155.1 | 2 | 958 | ||
XM_008765184.1 | XP_008763406.1 | 2 | 960 | |||
GenBank mRNA | BC088306.1 | - | 1 | 213 |
