siRNA Details
Gene Symbol: Lrrc39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109637.1 | NP_001103107.1 | 3 | 368 | ||
XM_006233231.2 | XP_006233293.1 | 3 | 355 | |||
GenBank mRNA | BC158606.1 | - | 3 | 324 | ||
FQ215425.1 | - | 3 | 309 |
