siRNA Details
Gene Symbol: Ppp2r5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764959.1 | XP_008763181.1 | 3 | 240 | ||
XM_008764960.1 | XP_008763182.1 | 3 | 240 | |||
XM_008764961.1 | XP_008763183.1 | 3 | 240 | |||
XM_008764962.1 | XP_008763184.1 | 3 | 240 |
