siRNA Details
Gene Symbol: Ppp2r5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191112.1 | NP_001178041.1 | 7 | 789 | ||
XM_006240559.2 | XP_006240621.1 | 7 | 812 | |||
XM_008764959.1 | XP_008763181.1 | 9 | 910 | |||
XM_008764960.1 | XP_008763182.1 | 9 | 910 | |||
XM_008764961.1 | XP_008763183.1 | 9 | 910 | |||
XM_008764962.1 | XP_008763184.1 | 9 | 910 | |||
XM_008764963.1 | XP_008763185.1 | 7 | 2289 |

More Information
Other Species Targeted

Gene Ontology Categories:
