siRNA Details
Gene Symbol: LOC100912557 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | FQ211017.1 | - | 2, 3 | 164 | ||
FQ217325.1 | - | 1, 2 | 118 | |||
FQ222783.1 | - | 2, 3 | 167 | |||
FQ224630.1 | - | 2, 3 | 146 |

More Information
Other Species Targeted

