siRNA Details
Gene Symbol: LOC100912557 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | FQ211017.1 | - | 4 | 306 | ||
FQ217325.1 | - | 3 | 260 | |||
FQ222783.1 | - | 4 | 309 | |||
FQ224630.1 | - | 4 | 288 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
