siRNA Details
Gene Symbol: Vom2r54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099494.1 | NP_001092964.1 | 2 | 675 | ||
XM_006241129.1 | XP_006241191.1 | 3 | 894 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
