siRNA Details
Gene Symbol: Tmem190 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001078306.4 | XP_001078306.2 | 5 | 1754 | ||
XM_008759018.1 | XP_008757240.1 | 5 | 1754 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
