siRNA Details
Gene Symbol: Slc25a54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109640.1 | NP_001103110.1 | 8 | 1135 | ||
XM_008761423.1 | XP_008759645.1 | 8 | 1156 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
