siRNA Details
Gene Symbol: Snapc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109643.2 | NP_001103113.1 | 2 | 191 | ||
GenBank mRNA | EV775546.1 | - | 2 | 227 |

More Information
Other Species Targeted

Gene Ontology Categories:
