siRNA Details
Gene Symbol: Ttc19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109644.1 | NP_001103114.1 | 6 | 630 | ||
XM_006246530.2 | XP_006246592.1 | 7 | 647 |

More Information
Other Species Targeted

Gene Ontology Categories:
