siRNA Details
Gene Symbol: Nop10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126100.1 | NP_001119572.1 | 2 | 232 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_025403, AK004120, AK012875, BC028497.
Gene Ontology Categories:
