siRNA Details
Gene Symbol: Katnbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109645.2 | NP_001103115.1 | 2, 3 | 206 | ||
XM_006234712.2 | XP_006234774.1 | 2, 3 | 259 | |||
GenBank mRNA | BC161951.1 | - | 2, 3 | 206 | 9026012 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_024254, BC062099, BC080661, AK036861, AK152570, and others.
