siRNA Details
Gene Symbol: Ubap1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006243297.1 | XP_006243359.1 | 2, 3 | 763 | ||
XM_006243298.1 | XP_006243360.1 | 3, 4 | 838 | |||
XM_006243299.2 | XP_006243361.1 | 2, 3 | 644 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
