siRNA Details
Gene Symbol: Acp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135562.1 | NP_001129034.1 | 3 | 292 | ||
XM_008764572.1 | XP_008762794.1 | 3 | 292 | |||
GenBank mRNA | BC158763.1 | - | 3 | 292 | 7109165 | |
BC161990.1 | - | 3 | 238 | 9032684 | ||
FQ215147.1 | - | 3 | 230 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
