siRNA Details
Gene Symbol: LOC691670 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109646.1 | NP_001103116.1 | 2 | 136 | ||
XM_008770702.1 | XP_008768924.1 | 2 | 189 | |||
XM_008770703.1 | XP_008768925.1 | 2 | 189 |
