siRNA Details
Gene Symbol: Tuba1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044270.2 | NP_001037735.1 | 4 | 1476 | ||
GenBank mRNA | BC060572.1 | - | 4 | 1480 | 6920989 | |
BC076379.1 | - | 4 | 1462 | 7106502 | ||
FQ212426.1 | - | 4 | 1471 | |||
FQ212740.1 | - | 4 | 1471 | |||
FQ213098.1 | - | 4 | 1476 | |||
FQ213226.1 | - | 4 | 1474 | |||
FQ213439.1 | - | 4 | 1473 | |||
FQ213947.1 | - | 4 | 1472 | |||
FQ214075.1 | - | 4 | 1474 | |||
FQ214100.1 | - | 4 | 1471 | |||
FQ214301.1 | - | 4 | 1475 | |||
FQ223081.1 | - | 4 | 1476 | |||
FQ225046.1 | - | 4 | 1480 | |||
FQ225339.1 | - | 4 | 1470 | |||
FQ225425.1 | - | 4 | 1476 | |||
FQ225650.1 | - | 4 | 1473 | |||
FQ228903.1 | - | 1259 | ||||
FQ231550.1 | - | 4 | 1472 | |||
FQ232160.1 | - | 3 | 1381 | |||
FQ232527.1 | - | 4 | 1476 | |||
FQ233000.1 | - | 4 | 1473 | |||
FQ233566.1 | - | 4 | 1474 | |||
FQ234135.1 | - | 4 | 1471 | |||
V01226.1 | - | 1 | 459 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
