siRNA Details
Gene Symbol: LOC691931 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109651.1 | NP_001103121.1 | 2 | 371 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_027482, BC116791, BC116793, AK012470, AK017758, and others.
