siRNA Details
Gene Symbol: Mlip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766417.1 | XP_008764639.1 | 2, 3 | 571 | ||
XM_008766420.1 | XP_008764642.1 | 2, 3 | 304 |

More Information
Other Species Targeted

Gene Ontology Categories:
