siRNA Details
Gene Symbol: Rps16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169146.1 | NP_001162617.1 | 5 | 387 | ||
XM_006228612.2 | XP_006228674.1 | 4 | 473 | |||
GenBank mRNA | BC064030.1 | - | 361 | |||
BC084715.1 | - | 5 | 356 | |||
BC159438.1 | - | 5 | 358 | |||
FQ211832.1 | - | 5 | 368 | |||
FQ221089.1 | - | 5 | 375 | |||
FQ221637.1 | - | 5 | 369 | |||
FQ221660.1 | - | 5 | 368 | |||
FQ221692.1 | - | 5 | 366 | |||
FQ223100.1 | - | 5 | 369 | |||
FQ223384.1 | - | 5 | 368 | |||
FQ223396.1 | - | 5 | 369 | |||
FQ224101.1 | - | 5 | 368 | |||
FQ228433.1 | - | 5 | 369 | |||
FQ229060.1 | - | 5 | 321 | |||
X17665.1 | - | 5 | 367 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
